Assay Details
| Gene Symbol: C1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031036.2 | NP_001026207.2 | 3 - 4 | 344 | 80 | |
| NM_001244737.1 | NP_001231666.1 | 2 - 3 | 466 | 80 | ||
| XM_015278043.2 | XP_015133529.1 | 2 - 3 | 227 | 80 | ||
| GenBank mRNA | AJ453246.1 | - | 3 - 4 | 345 | 80 | |
| AJ721061.1 | - | 3 - 4 | 344 | 80 | ||
| BU325589.1 | - | 2 - 3 | 435 | 80 | ||
| CV889988.1 | - | 3 - 4 | 341 | 80 | ||
| DR419424.1 | - | 415 | 80 | |||
More Information
Set Membership: |
|

C1D
Genomic Map