Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031114.1 | NP_001026285.1 | 1 - 2 | 132 | 66 | |
| XM_025149546.1 | XP_025005314.1 | 1 - 2 | 170 | 66 | ||
| GenBank mRNA | AJ720103.1 | - | 1 - 2 | 132 | 66 | |
More Information
Set Membership: |
|

CLIC2
Genomic Map