Assay Details
| Gene Symbol: DKC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031115.1 | NP_001026286.1 | 6 - 7 | 553 | 62 | |
| GenBank mRNA | AJ720455.1 | - | 6 - 7 | 553 | 62 | |
More Information
Set Membership: |
|

DKC1
Genomic Map