Assay Details
| Gene Symbol: USP12-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307997.3 | NP_001294926.1 | 3 - 4 | 451 | 129 | |
| GenBank mRNA | AJ851557.1 | - | 3 - 4 | 410 | 129 | |
More Information
Set Membership: |
|

USP12-like
Genomic Map