Assay Details
| Gene Symbol: USP12-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ851557.1 | - | 4 - 5 | 638 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP12-like| Gene Symbol: USP12-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ851557.1 | - | 4 - 5 | 638 | 143 | |
Set Membership: |
|