Assay Details
| Gene Symbol: USP12-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307997.3 | NP_001294926.1 | 6 - 7 | 844 | 69 | |
| XM_015278291.2 | XP_015133777.1 | 6 - 7 | 4841 | 69 | ||
| XM_015278292.2 | XP_015133778.1 | 4 - 5 | 465 | 69 | ||
| GenBank mRNA | AJ851557.1 | - | 7 - 8 | 889 | 69 | |
More Information
Set Membership: |
|

USP12-like
Genomic Map