Assay Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031129.1 | NP_001026300.1 | 1 - 2 | 178 | 142 | |
| XM_015285286.2 | XP_015140772.1 | 1 - 2 | 754 | 142 | ||
| GenBank mRNA | AJ719384.1 | - | 1 - 2 | 178 | 142 | |
More Information
Set Membership: |
|

CTSO
Genomic Map