Assay Details
| Gene Symbol: UFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031136.1 | NP_001026307.1 | 11 - 12 | 1347 | 77 | |
| XM_015276225.2 | XP_015131711.1 | 11 - 12 | 1369 | 77 | ||
| XM_015276226.2 | XP_015131712.1 | 11 - 12 | 1366 | 77 | ||
| GenBank mRNA | AJ720831.1 | - | 11 - 12 | 1347 | 77 | |
| BX933253.1 | - | 4 - 5 | 471 | 77 | ||
| BX934444.1 | - | 92 | 77 | |||
More Information
Set Membership: |
|

UFSP2
Genomic Map