Assay Details
| Gene Symbol: DNAJB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031224.1 | NP_001026395.1 | 5 - 6 | 728 | 61 | |
| XM_015288206.2 | XP_015143692.1 | 5 - 6 | 764 | 61 | ||
| GenBank mRNA | AJ851623.1 | - | 5 - 6 | 751 | 61 | |
More Information
Set Membership: |
|

DNAJB12
Genomic Map