Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031242.1 | NP_001026413.1 | 2 - 3 | 229 | 119 | |
| XM_015289156.2 | XP_015144642.1 | 2 - 3 | 460 | 119 | ||
| GenBank mRNA | AJ720295.1 | - | 2 - 3 | 229 | 119 | |
More Information
Set Membership: |
|

NEMP2
Genomic Map