Assay Details
| Gene Symbol: LANCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031245.2 | NP_001026416.2 | 2 - 3 | 230 | 65 | |
| GenBank mRNA | AJ720034.1 | - | 2 - 3 | 211 | 65 | |
More Information
Set Membership: |
|

LANCL1
Genomic Map