Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031249.1 | NP_001026420.1 | 5 - 6 | 766 | 81 | |
| XM_015289393.2 | XP_015144879.1 | 5 - 6 | 791 | 81 | ||
| XM_015289395.2 | XP_015144881.1 | 5 - 6 | 831 | 81 | ||
| XM_025152162.1 | XP_025007930.1 | 6 - 7 | 989 | 81 | ||
| GenBank mRNA | AJ719550.1 | - | 5 - 6 | 766 | 81 | |
More Information
Set Membership: |
|

STK17B
Genomic Map