Assay Details
| Gene Symbol: PSMD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031256.2 | NP_001026427.2 | 3 - 4 | 259 | 62 | |
| XM_025152195.1 | XP_025007963.1 | 1 - 2 | 55 | 62 | ||
| GenBank mRNA | AJ720599.1 | - | 3 - 4 | 226 | 62 | |
| CD734671.1 | - | 3 - 4 | 259 | 62 | ||
| CR387677.1 | - | 3 - 4 | 231 | 62 | ||
More Information
Set Membership: |
|

PSMD14
Genomic Map