Assay Details
| Gene Symbol: NEK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031264.1 | NP_001026435.1 | 2 - 3 | 289 | 64 | |
| XM_015290270.2 | XP_015145756.1 | 2 - 3 | 124 | 64 | ||
| XM_015290271.1 | XP_015145757.1 | 2 - 3 | 264 | 64 | ||
| XM_025152803.1 | XP_025008571.1 | 2 - 3 | 101 | 64 | ||
| GenBank mRNA | AJ851609.1 | - | 2 - 3 | 289 | 64 | |
More Information
Set Membership: |
|

NEK7
Genomic Map