Assay Details
| Gene Symbol: TRIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031431.1 | NP_001026602.1 | 3 - 4 | 521 | 69 | |
| GenBank mRNA | AJ720736.1 | - | 3 - 4 | 521 | 69 | |
More Information
Set Membership: |
|

TRIM14
Genomic Map