Assay Details
| Gene Symbol: PTGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031435.1 | NP_001026606.1 | 4 - 5 | 438 | 74 | |
| GenBank mRNA | AJ720800.1 | - | 4 - 5 | 438 | 74 | |
More Information
Set Membership: |
|

PTGR1
Genomic Map