Assay Details
| Gene Symbol: NUDT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031458.1 | NP_001026629.1 | 1 - 2 | 736 | 96 | |
| GenBank mRNA | AJ719921.1 | - | 1 - 2 | 736 | 96 | |
More Information
Set Membership: |
|

NUDT19
Genomic Map