Assay Details
| Gene Symbol: EARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031467.1 | NP_001026638.1 | 2 - 3 | 243 | 61 | |
| GenBank mRNA | AJ720568.1 | - | 2 - 3 | 243 | 61 | |
More Information
Set Membership: |
|

EARS2
Genomic Map