Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031499.1 | NP_001026670.1 | 223 | 98 | ||
| GenBank mRNA | AJ851744.1 | - | 223 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YOD1| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031499.1 | NP_001026670.1 | 223 | 98 | ||
| GenBank mRNA | AJ851744.1 | - | 223 | 98 | ||
Set Membership: |
|