Assay Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031505.1 | NP_001026676.1 | 4 - 5 | 523 | 63 | |
| XM_015282933.2 | XP_015138419.1 | 3 - 4 | 310 | 63 | ||
| GenBank mRNA | AJ719519.1 | - | 4 - 5 | 523 | 63 | |
More Information
Set Membership: |
|

PTDSS1
Genomic Map