Assay Details
| Gene Symbol: ADSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031521.1 | NP_001026692.1 | 9 - 10 | 965 | 70 | |
| XM_015283953.1 | XP_015139439.1 | 8 - 9 | 1075 | 70 | ||
| GenBank mRNA | AJ720419.1 | - | 9 - 10 | 965 | 70 | |
More Information
Set Membership: |
|

ADSS2
Genomic Map