Assay Details
| Gene Symbol: B3GAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031527.1 | NP_001026698.1 | 2 - 3 | 533 | 72 | |
| XM_025148810.1 | XP_025004578.1 | 1 - 2 | 780 | 72 | ||
| GenBank mRNA | AJ889606.1 | - | 2 - 3 | 533 | 72 | |
More Information
Set Membership: |
|

B3GAT2
Genomic Map