Assay Details
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031527.1 | NP_001026698.1 | 4 - 5 | 828 | 63 | |
XM_025148810.1 | XP_025004578.1 | 3 - 4 | 1075 | 63 | ||
GenBank mRNA | AJ889606.1 | - | 4 - 5 | 828 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
