Assay Details
| Gene Symbol: ETFDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031534.1 | NP_001026705.1 | 2 - 3 | 231 | 65 | |
| XM_015285303.2 | XP_015140789.2 | 2 - 3 | 224 | 65 | ||
| XM_025149748.1 | XP_025005516.1 | 2 - 3 | 231 | 65 | ||
| XM_025149749.1 | XP_025005517.1 | 2 - 3 | 336 | 65 | ||
| XM_025149750.1 | XP_025005518.1 | 2 - 3 | 170 | 65 | ||
| GenBank mRNA | AJ851712.1 | - | 2 - 3 | 231 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015285303.2, XM_025149750.1 transcripts.
Set Membership: |
|

ETFDH
Genomic Map