Assay Details
| Gene Symbol: AREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031537.1 | NP_001026708.1 | 4 - 5 | 617 | 121 | |
| XM_015276315.2 | XP_015131801.1 | 4 - 5 | 695 | 121 | ||
| GenBank mRNA | AY725836.1 | - | 4 - 5 | 617 | 121 | |
More Information
Set Membership: |
|

AREG
Genomic Map