Assay Details
| Gene Symbol: NPY4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031555.1 | NP_001026726.1 | 1 | 948 | 116 | |
| XM_015288399.2 | XP_015143885.1 | 2 | 2152 | 116 | ||
More Information
Set Membership: |
|

NPY4R
Genomic Map