Assay Details
| Gene Symbol: PGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031556.2 | NP_001026727.1 | 2 - 3 | 488 | 92 | |
| GenBank mRNA | AJ719703.1 | - | 2 - 3 | 489 | 92 | |
More Information
Set Membership: |
|

PGAM1
Genomic Map