Assay Details
| Gene Symbol: NPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031560.2 | NP_001026731.1 | 10 - 11 | 776 | 91 | |
| XM_015290325.2 | XP_015145811.1 | 10 - 11 | 798 | 91 | ||
| XM_025152895.1 | XP_025008663.1 | 9 - 10 | 996 | 91 | ||
| XM_025152896.1 | XP_025008664.1 | 9 - 10 | 757 | 91 | ||
| XM_025152897.1 | XP_025008665.1 | 9 - 10 | 724 | 91 | ||
| XM_025152898.1 | XP_025008666.1 | 10 - 11 | 1540 | 91 | ||
| GenBank mRNA | AJ720150.1 | - | 10 - 11 | 778 | 91 | |
More Information
Set Membership: |
|

NPL
Genomic Map