Assay Details
| Gene Symbol: CYR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031563.1 | NP_001026734.1 | 3 - 4 | 675 | 97 | |
| GenBank mRNA | J04496.1 | - | 3 - 4 | 675 | 97 | |
More Information
Set Membership: |
|

CYR61
Genomic Map