Assay Details
Gene Symbol: MCCC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031565.1 | NP_001026736.1 | 5 - 6 | 906 | 111 | |
XM_025152899.1 | XP_025008667.1 | 5 - 6 | 909 | 111 | ||
GenBank mRNA | AJ719309.1 | - | 5 - 6 | 906 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
