Assay Details
| Gene Symbol: GALNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031578.1 | NP_001026749.1 | 8 - 9 | 1790 | 61 | |
| XM_015300369.2 | XP_015155855.1 | 7 - 8 | 1503 | 61 | ||
| XM_025145312.1 | XP_025001080.1 | 8 - 9 | 1779 | 61 | ||
| GenBank mRNA | AJ851373.1 | - | 8 - 9 | 1790 | 61 | |
More Information
Set Membership: |
|

GALNT6
Genomic Map