Assay Details
| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031586.1 | NP_001026757.1 | 4 - 5 | 339 | 65 | |
| GenBank mRNA | AJ851632.1 | - | 4 - 5 | 339 | 65 | |
More Information
Set Membership: |
|

CDK5RAP3
Genomic Map