Assay Details
| Gene Symbol: C25H1ORF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031590.3 | NP_001026761.2 | 1 - 2 | 123 | 77 | |
| NM_001305796.2 | NP_001292725.1 | 1 - 2 | 123 | 77 | ||
| GenBank mRNA | AJ719894.1 | - | 1 - 2 | 125 | 77 | |
More Information
Set Membership: |
|

C25H1ORF43
Genomic Map