Assay Details
| Gene Symbol: PELO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031592.1 | NP_001026763.1 | 1 - 2 | 910 | 104 | |
| GenBank mRNA | AJ720283.1 | - | 1 - 2 | 928 | 104 | |
More Information
Set Membership: |
|

PELO
Genomic Map