Assay Details
| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031607.1 | NP_001026778.1 | 3 - 4 | 1030 | 107 | |
| XM_015293825.2 | XP_015149311.2 | 3 - 4 | 1133 | 107 | ||
| GenBank mRNA | AJ719651.1 | - | 3 - 4 | 1030 | 107 | |
More Information
Set Membership: |
|

N4BP3
Genomic Map