Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030645.1 | NP_001025816.1 | 2 - 3 | 423 | 66 | |
| GenBank mRNA | AJ720854.1 | - | 2 - 3 | 423 | 66 | |
More Information
Set Membership: |
|

NMRAL1
Genomic Map