Assay Details
| Gene Symbol: NRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030646.1 | NP_001025817.1 | 7 - 8 | 1020 | 71 | |
| XM_015282417.2 | XP_015137903.1 | 7 - 8 | 6330 | 71 | ||
| XM_015282467.2 | XP_015137953.1 | 7 - 8 | 6196 | 71 | ||
| XM_015282528.2 | XP_015138014.1 | 7 - 8 | 1134 | 71 | ||
| XM_015282586.2 | XP_015138072.1 | 7 - 8 | 6204 | 71 | ||
| XM_015282649.2 | XP_015138135.1 | 7 - 8 | 4805 | 71 | ||
| XM_015282685.2 | XP_015138171.1 | 7 - 8 | 6329 | 71 | ||
| XM_015282728.2 | XP_015138214.1 | 7 - 8 | 6329 | 71 | ||
| XM_015282769.2 | XP_015138255.1 | 7 - 8 | 6325 | 71 | ||
| XM_015282829.2 | XP_015138315.1 | 7 - 8 | 1038 | 71 | ||
| XM_025147172.1 | XP_025002940.1 | 6 - 7 | 1108 | 71 | ||
| GenBank mRNA | CR391678.1 | - | 4 - 5 | 522 | 71 | |
More Information
Set Membership: |
|

NRF1
Genomic Map