Assay Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270716.1 | NP_001257645.1 | 5 - 6 | 526 | 88 | |
| XM_015275319.2 | XP_015130805.1 | 5 - 6 | 531 | 88 | ||
| GenBank mRNA | AJ721085.1 | - | 4 - 5 | 903 | 88 | |
| BU296761.1 | - | 5 - 6 | 536 | 88 | ||
More Information
Set Membership: |
|

DERL3
Genomic Map