Assay Details
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030667.1 | NP_001025838.1 | 1 - 2 | 162 | 123 | |
| XM_015275320.2 | XP_015130806.1 | 1 - 2 | 144 | 123 | ||
| XM_015275321.2 | XP_015130807.1 | 1 - 2 | 144 | 123 | ||
| GenBank mRNA | AJ719424.1 | - | 1 - 2 | 162 | 123 | |
More Information
Set Membership: |
|

DDTL
Genomic Map