Assay Details
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030667.1 | NP_001025838.1 | 2 - 3 | 328 | 75 | |
| GenBank mRNA | AJ719424.1 | - | 2 - 3 | 328 | 75 | |
More Information
Set Membership: |
|

DDTL
Genomic Map