Assay Details
| Gene Symbol: MVB12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030677.1 | NP_001025848.1 | 11 - 12 | 1141 | 63 | |
| XM_015279389.2 | XP_015134875.1 | 9 - 10 | 995 | 63 | ||
| XM_015279390.2 | XP_015134876.1 | 10 - 11 | 2793 | 63 | ||
| XM_015279391.2 | XP_015134877.1 | 11 - 12 | 1112 | 63 | ||
| XM_015279392.2 | XP_015134878.1 | 10 - 11 | 1021 | 63 | ||
| XM_015279393.2 | XP_015134879.1 | 10 - 11 | 1126 | 63 | ||
| XM_015279394.2 | XP_015134880.1 | 9 - 10 | 802 | 63 | ||
| GenBank mRNA | AJ851783.1 | - | 11 - 12 | 1141 | 63 | |
More Information
Set Membership: |
|

MVB12B
Genomic Map