Assay Details
| Gene Symbol: PBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030678.1 | NP_001025849.1 | 2 - 3 | 432 | 73 | |
| XM_025156038.1 | XP_025011806.1 | 2 - 3 | 385 | 73 | ||
| XM_025156039.1 | XP_025011807.1 | 2 - 3 | 274 | 73 | ||
| XM_025156040.1 | XP_025011808.1 | 2 - 3 | 1176 | 73 | ||
| GenBank mRNA | AJ719751.1 | - | 2 - 3 | 432 | 73 | |
| BX931661.2 | - | 2 - 3 | 215 | 73 | ||
| GQ390248.1 | - | 2 - 3 | 90 | 73 | ||
More Information
Set Membership: |
|

PBX3
Genomic Map