Assay Details
| Gene Symbol: GBGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030683.1 | NP_001025854.1 | 1 - 2 | 152 | 69 | |
| GenBank mRNA | AJ719730.1 | - | 1 - 2 | 152 | 69 | |
More Information
Set Membership: |
|

GBGT1
Genomic Map