Assay Details
| Gene Symbol: TOR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030687.1 | NP_001025858.1 | 3 - 4 | 655 | 143 | |
| GenBank mRNA | AJ720743.1 | - | 3 - 4 | 655 | 143 | |
More Information
Set Membership: |
|

TOR1A
Genomic Map