Assay Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030689.2 | NP_001025860.2 | 1 - 2 | 100 | 150 | |
| GenBank mRNA | AJ719794.1 | - | 1 - 2 | 100 | 150 | |
| DR424106.1 | - | 1 - 2 | 77 | 150 | ||
More Information
Set Membership: |
|

DOLPP1
Genomic Map