Assay Details
| Gene Symbol: TLR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030693.1 | NP_001025864.1 | 2 - 3 | 274 | 160 | |
| GenBank mRNA | JQ711146.1 | - | 2 - 3 | 274 | 160 | |
| JQ711147.1 | - | 2 - 3 | 274 | 160 | ||
| JQ711148.1 | - | 2 - 3 | 274 | 160 | ||
| JQ711149.1 | - | 2 - 3 | 274 | 160 | ||
| JQ711150.1 | - | 2 - 3 | 274 | 160 | ||
| JQ711151.1 | - | 2 - 3 | 274 | 160 | ||
| JQ711152.1 | - | 2 - 3 | 274 | 160 | ||
| KF421483.1 | - | 2 - 3 | 274 | 160 | ||
| KF421484.1 | - | 2 - 3 | 274 | 160 | ||
| KF421485.1 | - | 2 - 3 | 274 | 160 | ||
| KF421486.1 | - | 2 - 3 | 274 | 160 | ||
| KP410249.1 | - | 2 - 3 | 274 | 160 | ||
More Information
Set Membership: |
|

TLR4
Genomic Map