Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030704.1 | NP_001025875.1 | 2 - 3 | 284 | 62 | |
| XM_015279872.1 | XP_015135358.1 | 4 - 5 | 592 | 62 | ||
| XM_025141598.1 | XP_024997366.1 | 4 - 5 | 358 | 62 | ||
| GenBank mRNA | AJ720170.1 | - | 2 - 3 | 284 | 62 | |
More Information
Set Membership: |
|

MMD
Genomic Map