Assay Details
| Gene Symbol: NCF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030709.1 | NP_001025880.1 | 3 - 4 | 279 | 53 | |
| XM_015295704.2 | XP_015151190.1 | 2 - 3 | 205 | 53 | ||
| GenBank mRNA | AJ719555.1 | - | 3 - 4 | 279 | 53 | |
More Information
Set Membership: |
|

NCF1C
Genomic Map