Assay Details
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030709.1 | NP_001025880.1 | 7 - 8 | 725 | 122 | |
XM_015295704.2 | XP_015151190.1 | 6 - 7 | 651 | 122 | ||
GenBank mRNA | AJ719555.1 | - | 7 - 8 | 725 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
