Assay Details
| Gene Symbol: RFFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030711.1 | NP_001025882.1 | 2 - 3 | 214 | 75 | |
| XM_015295722.2 | XP_015151208.1 | 2 - 3 | 219 | 75 | ||
| XM_015295723.2 | XP_015151209.1 | 2 - 3 | 279 | 75 | ||
| XM_015295725.2 | XP_015151211.1 | 2 - 3 | 290 | 75 | ||
| XM_025141948.1 | XP_024997716.1 | 3 - 4 | 1504 | 75 | ||
| XM_025141949.1 | XP_024997717.1 | 2 - 3 | 211 | 75 | ||
| XM_025141950.1 | XP_024997718.1 | 2 - 3 | 309 | 75 | ||
| GenBank mRNA | AJ720958.1 | - | 2 - 3 | 214 | 75 | |
More Information
Set Membership: |
|

RFFL
Genomic Map